Gene Transcripts
- Gene Symbol
- TRAPPC6B
- Entrez Gene ID
- 122553
- Gene Name
- trafficking protein particle complex 6B
- Gene Aliases
- TPC6
- Chromosome Location
- Chr. 14: 39147811 - 39170430 on Build GRCh38
- UniGene ID
- Hs.13303
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001079537.1 |
NP_001073005.1
|
1, 2 |
421 |
|
NM_177452.3 |
NP_803235.1
|
1, 2 |
421 |
|
XM_017020964.1 |
XP_016876453.1
|
1, 2 |
394 |
|
GenBank mRNA |
AK056690.1 |
- |
1, 2 |
391 |
|
AK310730.1 |
- |
1, 2 |
198 |
|
AK315900.1 |
- |
1, 2 |
316 |
|
AL833179.1 |
- |
1, 2 |
305 |
|
BC047328.1 |
- |
1, 2 |
305 |
5534151 |
BX161510.1 |
- |
1, 2 |
313 |
|
BX248013.1 |
- |
1, 2 |
344 |
|
View Full Product Details