Gene Transcripts

Gene Symbol
FBXL19-AS1
Entrez Gene ID
283932
Gene Name
FBXL19 antisense RNA 1 (head to head)
Gene Aliases
NCRNA00095
Chromosome Location
Chr. 16: 30919319 - 30923269 on Build GRCh38
UniGene ID
Hs.711535
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024348.1 1 618
GenBank mRNA AK092263.1 - 1 618
BC101464.1 - 1 544
BC101988.1 - 1 544
BC105687.1 - 1 544
BC105926.1 - 1 544

View Full Product Details