Gene Transcripts

Gene Symbol
PARP6
Entrez Gene ID
56965
Gene Name
poly(ADP-ribose) polymerase family member 6
Gene Aliases
ARTD17, PARP-6-B1, PARP-6-C, pART17
Chromosome Location
Chr. 15: 72241181 - 72272575 on Build GRCh38
UniGene ID
Hs.270244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323515.1 NP_001310444.1 5 584
NM_001323516.1 NP_001310445.1 5 657
NM_001323519.1 NP_001310448.1 5 706
NM_001323521.1 NP_001310450.1 5 651
NM_001323522.1 NP_001310451.1 5 596
NM_001323523.1 NP_001310452.1 5 879
NM_001323524.1 NP_001310453.1 5 657
NM_001323525.1 NP_001310454.1 5 767
NM_001323526.1 NP_001310455.1 5 767
NM_001323528.1 NP_001310457.1 5 657
NM_001323530.1 NP_001310459.1 5 767
NM_001323531.1 NP_001310460.1 5 596
NM_001323532.1 NP_001310461.1 5 767
NM_020214.3 NP_064599.2 5 657
NR_136594.1 5 767
NR_136596.1 5 767
NR_136599.1 5 657
NR_136603.1 5 651
NR_136604.1 5 767
NR_136605.1 5 767
NR_136606.1 5 879
NR_136607.1 5 767
NR_136608.1 5 657
NR_136609.1 5 767
NR_136610.1 5 767
NR_136611.1 5 767
XM_017022418.1 XP_016877907.1 4 1164
GenBank mRNA AB499727.1 - 2 125
AB499728.1 - 2 125
AK021790.1 - 3 219
AK022547.1 - 4 348
AK126416.1 - 2 950
AL390087.1 - 2 328
BC110902.1 - 3 173 6194444
BC119688.2 - 3 184
BC119689.1 - 3 184

View Full Product Details