Gene Transcripts
- Gene Symbol
- NLRX1
- Entrez Gene ID
- 79671
- Gene Name
- NLR family member X1
- Gene Aliases
- CLR11.3, DLNB26, NOD26, NOD5, NOD9
- Chromosome Location
- Chr. 11: 119168334 - 119184016 on Build GRCh38
- UniGene ID
- Hs.524082
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282143.1 |
NP_001269072.1
|
6, 7 |
1909 |
|
NM_001282144.1 |
NP_001269073.1
|
6, 7 |
2317 |
|
NM_001282358.1 |
NP_001269287.1
|
6, 7 |
1841 |
|
NM_024618.3 |
NP_078894.2
|
6, 7 |
1936 |
|
XM_005271669.1 |
XP_005271726.1
|
6, 7 |
2160 |
|
XM_005271670.1 |
XP_005271727.1
|
5, 6 |
1813 |
|
XM_005271671.1 |
XP_005271728.1
|
5, 6 |
1987 |
|
XM_005271672.1 |
XP_005271729.1
|
5, 6 |
1810 |
|
XM_006718904.1 |
XP_006718967.1
|
6, 7 |
2194 |
|
XM_011542980.1 |
XP_011541282.1
|
6, 7 |
1824 |
|
GenBank mRNA |
AB094095.1 |
- |
6, 7 |
1920 |
|
AK025131.1 |
- |
1, 2 |
563 |
|
AK056454.1 |
- |
5, 6 |
1634 |
|
AK095247.1 |
- |
4, 5 |
1121 |
|
AK225223.1 |
- |
1, 2 |
563 |
|
AK291716.1 |
- |
6, 7 |
1903 |
|
AK303002.1 |
- |
5, 6 |
1810 |
|
AY245437.1 |
- |
5, 6 |
1679 |
|
BC013199.1 |
- |
2, 3 |
1040 |
|
BC110890.1 |
- |
6, 7 |
2282 |
4937963 |
BX647705.1 |
- |
6, 7 |
1894 |
|
EF452237.1 |
- |
5, 6 |
1679 |
|
View Full Product Details