Gene Transcripts
- Gene Symbol
- RBM19
- Entrez Gene ID
- 9904
- Gene Name
- RNA binding motif protein 19
- Gene Aliases
- Chromosome Location
- Chr. 12: 113816738 - 113966371 on Build GRCh38
- UniGene ID
- Hs.7482
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001146698.1 |
NP_001140170.1
|
3, 4 |
492 |
|
NM_001146699.1 |
NP_001140171.1
|
3, 4 |
492 |
|
NM_016196.3 |
NP_057280.2
|
3, 4 |
492 |
|
XM_017020280.1 |
XP_016875769.1
|
3, 4 |
492 |
|
XM_017020281.1 |
XP_016875770.1
|
3, 4 |
492 |
|
GenBank mRNA |
AB014582.1 |
- |
3, 4 |
427 |
|
AK291444.1 |
- |
3, 4 |
443 |
|
AK314606.1 |
- |
3, 4 |
492 |
|
AL117547.1 |
- |
3, 4 |
433 |
|
BC004289.1 |
- |
3, 4 |
431 |
3619715 |
BC006137.1 |
- |
3, 4 |
431 |
3677625 |
DB096676.1 |
- |
3, 4 |
492 |
|
View Full Product Details