Gene Transcripts

Gene Symbol
NLRP12
Entrez Gene ID
91662
Gene Name
NLR family pyrin domain containing 12
Gene Aliases
CLR19.3, FCAS2, NALP12, PAN6, PYPAF7, RNO, RNO2
Chromosome Location
Chr. 19: 53793584 - 53824403 on Build GRCh38
UniGene ID
Hs.631573
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277126.1 NP_001264055.1 5 2561
NM_001277129.1 NP_001264058.1 5 2558
NM_144687.3 NP_653288.1 5 2558
XM_011527479.1 XP_011525781.1 5 2561
XM_011527480.1 XP_011525782.1 5 2561
XM_011527482.1 XP_011525784.1 5 2561
XM_017027460.1 XP_016882949.1 5 2561
XM_017027461.1 XP_016882950.1 5 2561
XM_017027462.1 XP_016882951.1 4 2387
XM_017027463.1 XP_016882952.1 6 2334
XM_017027464.1 XP_016882953.1 5 2132
XM_017027465.1 XP_016882954.1 6 2342
XM_017027466.1 XP_016882955.1 6 2339
XM_017027467.1 XP_016882956.1 5 2134
GenBank mRNA AF231021.1 - 3 1486
AK095460.1 - 5 2473
AK290772.1 - 5 2340
AY095146.1 - 5 2505
AY116204.1 - 5 2552
AY116205.1 - 5 2552
AY116206.1 - 5 2552
AY116207.1 - 5 2552
AY154467.1 - 5 2505
BC028069.1 - 5 2448 5212737

View Full Product Details