Gene Transcripts
- Gene Symbol
- NCAPH2
- Entrez Gene ID
- 29781
- Gene Name
- non-SMC condensin II complex subunit H2
- Gene Aliases
- CAPH2
- Chromosome Location
- Chr. 22: 50508216 - 50524780 on Build GRCh38
- UniGene ID
- Hs.730607
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001185011.1 |
NP_001171940.1
|
4, 5 |
466 |
|
NM_014551.4 |
NP_055366.3
|
4, 5 |
466 |
|
NM_152299.3 |
NP_689512.2
|
4, 5 |
466 |
|
XM_005261912.4 |
XP_005261969.1
|
5, 6 |
518 |
|
XM_011530685.2 |
XP_011528987.1
|
4, 5 |
346 |
|
XM_017028793.1 |
XP_016884282.1
|
5, 6 |
504 |
|
GenBank mRNA |
AL021682.1 |
- |
4, 5 |
435 |
|
BC000473.2 |
- |
4, 5 |
439 |
|
BC001509.2 |
- |
4, 5 |
439 |
|
BC001833.2 |
- |
4, 5 |
439 |
|
BC001937.1 |
- |
4, 5 |
423 |
2819717 |
BC009441.1 |
- |
4, 5 |
422 |
3509636 |
BC014939.1 |
- |
2, 3 |
126 |
|
CR456604.1 |
- |
4, 5 |
398 |
|
DB498781.1 |
- |
4, 5 |
467 |
|
View Full Product Details