Gene Transcripts
- Gene Symbol
- KNL1
- Entrez Gene ID
- 57082
- Gene Name
- kinetochore scaffold 1
- Gene Aliases
- AF15Q14, CASC5, CT29, D40, MCPH4, PPP1R55, Spc7, hKNL-1, hSpc105
- Chromosome Location
- Chr. 15: 40594012 - 40664342 on Build GRCh38
- UniGene ID
- Hs.181855
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144508.4 |
NP_653091.3
|
5, 6 |
356 |
|
NM_170589.4 |
NP_733468.3
|
5, 6 |
356 |
|
XM_017022432.1 |
XP_016877921.1
|
5, 6 |
593 |
|
GenBank mRNA |
AB022190.1 |
- |
5, 6 |
357 |
|
AF173994.1 |
- |
5, 6 |
585 |
|
AF248041.1 |
- |
5, 6 |
340 |
|
AF461041.1 |
- |
5, 6 |
340 |
|
AK307605.1 |
- |
5, 6 |
348 |
|
AK307996.1 |
- |
5, 6 |
346 |
|
BC029373.1 |
- |
5, 6 |
333 |
|
View Full Product Details