Gene Transcripts

Gene Symbol
RPL39
Entrez Gene ID
6170
Gene Name
ribosomal protein L39
Gene Aliases
L39, RPL39P42, RPL39_23_1806
Chromosome Location
Chr. X: 119786504 - 119791659 on Build GRCh38
UniGene ID
Hs.300141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000.3 NP_000991.1 2 166
GenBank mRNA AK098280.1 - 1 1664
BC001019.2 - 1 118 3343512
BC070205.1 - 1 136 6732169
BX476041.1 - 2 167
D79205.1 - 2 120
U57846.1 - 2 101

Gene Symbol
LMNTD1
Entrez Gene ID
160492
Gene Name
lamin tail domain containing 1
Gene Aliases
IFLTD1, PAS1C1
Chromosome Location
Chr. 12: 25476082 - 25648604 on Build GRCh38
UniGene ID
Hs.44647
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA DW009862.1 - 5 580

Gene Symbol
RPL39P5
Entrez Gene ID
553117
Gene Name
ribosomal protein L39 pseudogene 5
Gene Aliases
RPL39_7_406
Chromosome Location
Chr. 3: 134351739 - 134352143 on Build GRCh38
UniGene ID
Hs.611241
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB209077.1 - 2 295

View Full Product Details