Gene Transcripts
- Gene Symbol
- FLT3LG
- Entrez Gene ID
- 2323
- Gene Name
- fms related tyrosine kinase 3 ligand
- Gene Aliases
- FL, FLT3L
- Chromosome Location
- Chr. 19: 49474172 - 49487038 on Build GRCh38
- UniGene ID
- Hs.428
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001204502.1 |
NP_001191431.1
|
3, 4 |
259 |
|
NM_001204503.1 |
NP_001191432.1
|
2, 3 |
228 |
|
NM_001278638.1 |
NP_001265567.1
|
3, 4 |
276 |
|
NM_001459.3 |
NP_001450.2
|
3, 4 |
259 |
|
XM_005258680.4 |
XP_005258737.3
|
2, 3 |
249 |
|
XM_005258681.4 |
XP_005258738.3
|
2, 3 |
249 |
|
XM_005258682.4 |
XP_005258739.3
|
2, 3 |
250 |
|
XM_005258683.4 |
XP_005258740.3
|
2, 3 |
249 |
|
XM_006723116.3 |
XP_006723179.2
|
2, 3 |
250 |
|
XM_011526675.2 |
XP_011524977.1
|
3, 4 |
295 |
|
XM_011526676.2 |
XP_011524978.1
|
3, 4 |
260 |
|
XM_011526677.2 |
XP_011524979.1
|
2, 3 |
249 |
|
XM_011526678.2 |
XP_011524980.1
|
3, 4 |
295 |
|
XM_011526680.2 |
XP_011524982.1
|
2, 3 |
249 |
|
XM_011526682.2 |
XP_011524984.1
|
2, 3 |
266 |
|
XM_017026532.1 |
XP_016882021.1
|
2, 3 |
250 |
|
GenBank mRNA |
AI792400.1 |
- |
3, 4 |
240 |
|
AK301136.1 |
- |
2, 3 |
220 |
|
BC006331.2 |
- |
3, 4 |
249 |
|
BC011914.2 |
- |
3, 4 |
249 |
|
BC126293.1 |
- |
3, 4 |
216 |
8992009 |
BC136464.1 |
- |
3, 4 |
216 |
9020453 |
BC144129.1 |
- |
3, 4 |
216 |
9052649 |
U03858.1 |
- |
2, 3 |
232 |
|
U04806.1 |
- |
3, 4 |
241 |
|
View Full Product Details