Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002333.3 | NP_002324.2 | 5 | 606 | |
XM_005258945.1 | XP_005259002.1 | 5 | 523 | ||
GenBank mRNA | AB009462.1 | - | 5 | 739 | |
AK074751.1 | - | 3 | 322 | ||
AK126750.1 | - | 1 | 1414 | ||
AK129963.1 | - | 3 | 772 | ||
AK295505.1 | - | 3 | 827 | ||
AK296536.1 | - | 5 | 487 | ||
AK316314.1 | - | 4 | 433 | ||
BC007408.2 | - | 5 | 535 | 3009711 |