Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001257102.1 | NP_001244031.1 | 3 | 423 | |
NM_002491.2 | NP_002482.1 | 2 | 400 | ||
XM_011511230.2 | XP_011509532.1 | 3 | 437 | ||
XM_017004186.1 | XP_016859675.1 | 2 | 645 | ||
GenBank mRNA | AF035839.1 | - | 2 | 225 | |
AF047183.1 | - | 2 | 329 | ||
BC018183.1 | - | 2 | 348 | 3881592 | |
BM848639.1 | - | 2 | 400 | ||
BU188560.1 | - | 2 | 127 | ||
CR456924.1 | - | 1 | 77 |