Gene Transcripts

Gene Symbol
CSF2
Entrez Gene ID
1437
Gene Name
colony stimulating factor 2
Gene Aliases
GMCSF
Chromosome Location
Chr. 5: 132073792 - 132076170 on Build GRCh38
UniGene ID
Hs.1349
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000758.3 NP_000749.2 2 209
GenBank mRNA AF510855.1 - 2 126
AY720441.1 - 2 186
BC108724.1 - 2 196 3886571
BC113924.1 - 2 200 40084800
BC113999.1 - 2 200 40084798
KP271009.1 - 2 126
M10663.1 - 2 185
M11220.1 - 2 209
M11734.1 - 2 192

View Full Product Details