Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000980.3 | NP_000971.1 | 2 | 243 | |
GenBank mRNA | AK222647.1 | - | 2 | 199 | |
AK297323.1 | - | 2 | 200 | ||
AK297358.1 | - | 2 | 198 | ||
AK300809.1 | - | 1 | 195 | ||
AK311774.1 | - | 2 | 199 | ||
BC007512.2 | - | 1 | 183 | 2961519 | |
BC066319.1 | - | 1 | 203 | 5286436 | |
BC071920.1 | - | 1 | 183 | 6293838 | |
BI753938.1 | - | 186 | |||
HY050156.1 | - | 2 | 244 | ||
L05093.1 | - | 2 | 183 | ||
X80822.1 | - | 1 | 190 |