Gene Transcripts

Gene Symbol
HIST1H4D
Entrez Gene ID
8360
Gene Name
histone cluster 1, H4d
Gene Aliases
H4/b, H4FB, dJ221C16.9
Chromosome Location
Chr. 6: 26188710 - 26189076 on Build GRCh38
UniGene ID
Hs.248179
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003539.3 NP_003530.1 1 180
GenBank mRNA BC128104.1 - 1 180
BC128105.1 - 1 179
BC143045.1 - 1 180 40112087

View Full Product Details