Gene Transcripts

Gene Symbol
NRG2
Entrez Gene ID
9542
Gene Name
neuregulin 2
Gene Aliases
DON1, HRG2, NTAK
Chromosome Location
Chr. 5: 139846779 - 140043675 on Build GRCh38
UniGene ID
Hs.408515
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184935.1 NP_001171864.1 1 390
NM_004883.2 NP_004874.1 1 390
NM_013981.3 NP_053584.1 1 390
NM_013982.2 NP_053585.1 1 390
NM_013983.2 NP_053586.1 1 390
XM_006714810.3 XP_006714873.1 1 756
XM_011537713.2 XP_011536015.1 1 766
XM_017010066.1 XP_016865555.1 1 755
GenBank mRNA AB005060.1 - 1 385
U92987.1 - 1 312

View Full Product Details