Gene Transcripts
- Gene Symbol
- NTN3
- Entrez Gene ID
- 4917
- Gene Name
- netrin 3
- Gene Aliases
- NTN2L
- Chromosome Location
- Chr. 16: 2471499 - 2474145 on Build GRCh38
- UniGene ID
- Hs.158336
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006181.2 |
NP_006172.1
|
5 |
1563 |
|
GenBank mRNA |
U86759.1 |
- |
5 |
1563 |
|
View Full Product Details