Gene Transcripts

Gene Symbol
VPS28
Entrez Gene ID
51160
Gene Name
VPS28, ESCRT-I subunit
Gene Aliases
Chromosome Location
Chr. 8: 144423601 - 144428563 on Build GRCh38
UniGene ID
Hs.418175
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016208.3 NP_057292.1 4, 5 213
NM_183057.2 NP_898880.1 4, 5 213
XM_005272323.3 XP_005272380.1 4, 5 205
XM_005272324.3 XP_005272381.1 4, 5 205
GenBank mRNA AF182844.1 - 3, 4 165
AF316887.1 - 3, 4 147
AK026865.1 - 4, 5 163
BC006485.1 - 3, 4 149 3535787
BC019321.1 - 3, 4 149 2822758
BC050713.2 - 4, 5 161 5741129
BU658837.1 - 4, 5 506
KU178635.1 - 3, 4 104
KU178636.1 - 3, 4 104
R72904.1 - 3, 4 95

View Full Product Details