Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014485.2 | NP_055300.1 | 3 | 290 | |
GenBank mRNA | AK290075.1 | - | 3 | 290 | |
BC020734.1 | - | 3 | 267 | 4731411 | |
CB988786.1 | - | 317 | |||
CR541662.1 | - | 2 | 199 | ||
CR541679.1 | - | 2 | 199 | ||
D82073.1 | - | 2 | 209 |