Gene Transcripts

Gene Symbol
HIST1H1C
Entrez Gene ID
3006
Gene Name
histone cluster 1, H1c
Gene Aliases
H1.2, H1C, H1F2, H1s-1
Chromosome Location
Chr. 6: 26055740 - 26056471 on Build GRCh38
UniGene ID
Hs.7644
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005319.3 NP_005310.1 1 117
GenBank mRNA AB451259.1 - 1 74
AB451385.1 - 1 74
AK290947.1 - 1 116
BC002649.1 - 1 118

View Full Product Details