Gene Transcripts

Gene Symbol
IL26
Entrez Gene ID
55801
Gene Name
interleukin 26
Gene Aliases
AK155, IL-26
Chromosome Location
Chr. 12: 68201349 - 68225791 on Build GRCh38
UniGene ID
Hs.272350
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018402.1 NP_060872.1 1 109
GenBank mRNA AJ251549.1 - 1 109
BC066270.1 - 1 108 7002121
BC066271.1 - 1 108 7002122

View Full Product Details