Gene Transcripts

Gene Symbol
ZBTB47
Entrez Gene ID
92999
Gene Name
zinc finger and BTB domain containing 47
Gene Aliases
ZNF651
Chromosome Location
Chr. 3: 42653684 - 42667580 on Build GRCh38
UniGene ID
Hs.409561
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145166.3 NP_660149.2 2 1606
GenBank mRNA AK127065.1 - 3 1300
AL133062.1 - 1 299
BC021855.1 - 1 264 4943271

View Full Product Details