Gene Transcripts

Gene Symbol
Hdc
Entrez Gene ID
24443
Gene Name
histidine decarboxylase
Gene Aliases
Chromosome Location
Chr. 3: 119057524 - 119075599 on Build Rnor_6.0
UniGene ID
Rn.48653
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017016.2 NP_058712.2 2 211
GenBank mRNA BC128724.1 - 2 211
M29591.1 - 2 218

View Full Product Details