Gene Transcripts
- Gene Symbol
- Nol3
- Entrez Gene ID
- 85383
- Gene Name
- nucleolar protein 3 (apoptosis repressor with CARD domain)
- Gene Aliases
- Arc
- Chromosome Location
- Chr. 19: 37232482 - 37236683 on Build Rnor_6.0
- UniGene ID
- Rn.86956
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053516.2 |
NP_445968.2
|
2 |
709 |
|
GenBank mRNA |
U40627.1 |
- |
2 |
709 |
|
View Full Product Details