Gene Transcripts

Gene Symbol
Il18bp
Entrez Gene ID
84388
Gene Name
interleukin 18 binding protein
Gene Aliases
Igifbp
Chromosome Location
Chr. 1: 167091521 - 167095718 on Build Rnor_6.0
UniGene ID
Rn.20083
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053374.2 NP_445826.2 3 409
XM_008759805.1 XP_008758027.1 3 2567
GenBank mRNA AF154569.1 - 2 220

View Full Product Details