Gene Transcripts

Gene Symbol
VDAC1
Entrez Gene ID
7416
Gene Name
voltage dependent anion channel 1
Gene Aliases
PORIN, VDAC-1
Chromosome Location
Chr. 5: 133971875 - 134070987 on Build GRCh38
UniGene ID
Hs.519320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003374.2 NP_003365.1 9 1268
NR_036624.1 9 1346
NR_036625.1 9 1349
XM_005272075.3 XP_005272132.1 9 1265
XM_017009821.1 XP_016865310.1 9 1196
XM_017009822.1 XP_016865311.1 9 1175
XM_017009823.1 XP_016865312.1 9 1198
GenBank mRNA AK095989.1 - 9 1268
AK122953.1 - 12 2671
AK293639.1 - 7 725
BC008482.1 - 9 1053 4332199
BC071168.1 - 9 1085 6023095
BC090042.1 - 9 1078 30395350
L06132.1 - 9 1122

View Full Product Details