Gene Transcripts

Gene Symbol
Slc1a5
Entrez Gene ID
292657
Gene Name
solute carrier family 1 (neutral amino acid transporter), member 5
Gene Aliases
Asct2, H4-ASCT2, Slc1a7
Chromosome Location
Chr. 1: 78710686 - 78724789 on Build Rnor_6.0
UniGene ID
Rn.28965
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175758.3 NP_786934.1 1 934
GenBank mRNA AJ132846.1 - 1 665
BC080242.1 - 1 934 7100054

View Full Product Details