Gene Transcripts

Gene Symbol
Chgb
Entrez Gene ID
24259
Gene Name
chromogranin B (secretogranin 1)
Gene Aliases
Chromosome Location
Chr. 3: 125428260 - 125441601 on Build Rnor_6.0
UniGene ID
Rn.11090
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012526.2 NP_036658.2 2 348
XM_006235072.2 XP_006235134.1 3 258
GenBank mRNA AF019974.1 - 2 148
BC128743.1 - 2 167 8370781

View Full Product Details