Gene Transcripts

Gene Symbol
Pthlh
Entrez Gene ID
24695
Gene Name
parathyroid hormone-like hormone
Gene Aliases
PLP
Chromosome Location
Chr. 4: 181663425 - 181674181 on Build Rnor_6.0
UniGene ID
Rn.9750
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012636.1 NP_036768.1 223
GenBank mRNA M21967.1 - 149
M31603.1 - 223

View Full Product Details