Gene Transcripts
- Gene Symbol
- Tnnt2
- Entrez Gene ID
- 24837
- Gene Name
- troponin T type 2 (cardiac)
- Gene Aliases
- CTTG, Ctt, RATCTTG, Tnnt3
- Chromosome Location
- Chr. 13: 52662973 - 52680992 on Build Rnor_6.0
- UniGene ID
- Rn.9965
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012676.1 |
NP_036808.1
|
4, 5 |
161 |
|
XM_006249822.1 |
XP_006249884.1
|
6, 7 |
270 |
|
XM_006249823.1 |
XP_006249885.1
|
6, 7 |
271 |
|
XM_006249825.2 |
XP_006249887.1
|
6, 7 |
284 |
|
XM_006249828.1 |
XP_006249890.1
|
5, 6 |
258 |
|
XM_006249829.1 |
XP_006249891.1
|
5, 6 |
259 |
|
XM_006249833.2 |
XP_006249895.1
|
5, 6 |
266 |
|
XM_006249835.1 |
XP_006249897.1
|
4, 5 |
228 |
|
XM_006249836.1 |
XP_006249898.1
|
4, 5 |
229 |
|
XM_006249838.1 |
XP_006249900.1
|
4, 5 |
229 |
|
XM_006249840.2 |
XP_006249902.1
|
7, 8 |
300 |
|
XM_006249842.2 |
XP_006249904.1
|
5, 6 |
257 |
|
XM_008769535.1 |
XP_008767757.1
|
6, 7 |
271 |
|
XM_008769536.1 |
XP_008767758.1
|
6, 7 |
271 |
|
XM_008769537.1 |
XP_008767759.1
|
5, 6 |
259 |
|
XM_008769538.1 |
XP_008767760.1
|
4, 5 |
229 |
|
GenBank mRNA |
BC161855.1 |
- |
5, 6 |
266 |
|
EU295527.1 |
- |
3, 4 |
131 |
|
M26051.1 |
- |
4, 5 |
161 |
|
M26052.1 |
- |
4, 5 |
181 |
|
View Full Product Details