Gene Transcripts
- Gene Symbol
- Sparc
- Entrez Gene ID
- 24791
- Gene Name
- secreted protein, acidic, cysteine-rich (osteonectin)
- Gene Aliases
- Chromosome Location
- Chr. 10: 40742390 - 40764222 on Build Rnor_6.0
- UniGene ID
- Rn.234316
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012656.1 |
NP_036788.1
|
6, 7 |
383 |
|
XM_008767663.1 |
XP_008765885.1
|
5, 6 |
461 |
|
GenBank mRNA |
BC061777.1 |
- |
5, 6 |
398 |
|
D28875.1 |
- |
5, 6 |
400 |
|
FQ212364.1 |
- |
5, 6 |
452 |
|
FQ212499.1 |
- |
5, 6 |
417 |
|
FQ213815.1 |
- |
5, 6 |
417 |
|
FQ213945.1 |
- |
5, 6 |
417 |
|
FQ214222.1 |
- |
5, 6 |
415 |
|
FQ214492.1 |
- |
5, 6 |
417 |
|
FQ214649.1 |
- |
5, 6 |
417 |
|
FQ214673.1 |
- |
5, 6 |
417 |
|
FQ215195.1 |
- |
5, 6 |
416 |
|
FQ215581.1 |
- |
5, 6 |
418 |
|
FQ215622.1 |
- |
5, 6 |
419 |
|
FQ215818.1 |
- |
5, 6 |
428 |
|
FQ215935.1 |
- |
5, 6 |
417 |
|
FQ216823.1 |
- |
5, 6 |
419 |
|
FQ217546.1 |
- |
5, 6 |
418 |
|
FQ219932.1 |
- |
6, 7 |
399 |
|
FQ219983.1 |
- |
5, 6 |
417 |
|
FQ220003.1 |
- |
5, 6 |
417 |
|
FQ220043.1 |
- |
5, 6 |
415 |
|
FQ220046.1 |
- |
5, 6 |
417 |
|
FQ220186.1 |
- |
5, 6 |
415 |
|
FQ220279.1 |
- |
3, 4 |
241 |
|
FQ220287.1 |
- |
5, 6 |
419 |
|
FQ220443.1 |
- |
5, 6 |
399 |
|
FQ220507.1 |
- |
5, 6 |
425 |
|
FQ220547.1 |
- |
5, 6 |
417 |
|
FQ220572.1 |
- |
5, 6 |
417 |
|
FQ220644.1 |
- |
5, 6 |
417 |
|
FQ220827.1 |
- |
5, 6 |
417 |
|
FQ220880.1 |
- |
5, 6 |
417 |
|
FQ220901.1 |
- |
5, 6 |
417 |
|
FQ222224.1 |
- |
5, 6 |
417 |
|
FQ223025.1 |
- |
5, 6 |
417 |
|
FQ224996.1 |
- |
4, 5 |
288 |
|
FQ225033.1 |
- |
5, 6 |
419 |
|
FQ225092.1 |
- |
5, 6 |
413 |
|
FQ225200.1 |
- |
5, 6 |
417 |
|
FQ225220.1 |
- |
5, 6 |
396 |
|
FQ225486.1 |
- |
5, 6 |
399 |
|
FQ225609.1 |
- |
5, 6 |
417 |
|
FQ225618.1 |
- |
5, 6 |
415 |
|
FQ225644.1 |
- |
1, 2 |
84 |
|
FQ225678.1 |
- |
5, 6 |
417 |
|
FQ225688.1 |
- |
5, 6 |
417 |
|
FQ225694.1 |
- |
5, 6 |
419 |
|
FQ225722.1 |
- |
5, 6 |
415 |
|
FQ225884.1 |
- |
5, 6 |
415 |
|
FQ225913.1 |
- |
5, 6 |
417 |
|
FQ225914.1 |
- |
5, 6 |
419 |
|
FQ225965.1 |
- |
5, 6 |
417 |
|
FQ226299.1 |
- |
5, 6 |
399 |
|
FQ226354.1 |
- |
5, 6 |
417 |
|
FQ226360.1 |
- |
5, 6 |
417 |
|
FQ226584.1 |
- |
5, 6 |
417 |
|
FQ226599.1 |
- |
5, 6 |
418 |
|
FQ226651.1 |
- |
5, 6 |
440 |
|
FQ226673.1 |
- |
5, 6 |
398 |
|
FQ226731.1 |
- |
5, 6 |
417 |
|
FQ226746.1 |
- |
5, 6 |
415 |
|
FQ227279.1 |
- |
5, 6 |
417 |
|
FQ227628.1 |
- |
5, 6 |
425 |
|
FQ227644.1 |
- |
5, 6 |
417 |
|
FQ227708.1 |
- |
5, 6 |
417 |
|
FQ227725.1 |
- |
5, 6 |
417 |
|
FQ227727.1 |
- |
5, 6 |
396 |
|
FQ227749.1 |
- |
5, 6 |
417 |
|
FQ228032.1 |
- |
5, 6 |
397 |
|
FQ228597.1 |
- |
5, 6 |
399 |
|
FQ229172.1 |
- |
5, 6 |
417 |
|
FQ229199.1 |
- |
5, 6 |
417 |
|
FQ229230.1 |
- |
5, 6 |
417 |
|
FQ229283.1 |
- |
5, 6 |
399 |
|
FQ229299.1 |
- |
5, 6 |
400 |
|
FQ229315.1 |
- |
5, 6 |
415 |
|
FQ229316.1 |
- |
5, 6 |
417 |
|
FQ230029.1 |
- |
5, 6 |
417 |
|
FQ230086.1 |
- |
5, 6 |
415 |
|
FQ230381.1 |
- |
5, 6 |
417 |
|
FQ230595.1 |
- |
5, 6 |
399 |
|
FQ230619.1 |
- |
5, 6 |
417 |
|
FQ230667.1 |
- |
5, 6 |
417 |
|
FQ230705.1 |
- |
5, 6 |
415 |
|
FQ230778.1 |
- |
5, 6 |
417 |
|
FQ230864.1 |
- |
5, 6 |
417 |
|
FQ230993.1 |
- |
5, 6 |
417 |
|
FQ231067.1 |
- |
5, 6 |
417 |
|
FQ231166.1 |
- |
5, 6 |
416 |
|
FQ231444.1 |
- |
5, 6 |
416 |
|
FQ231523.1 |
- |
5, 6 |
417 |
|
FQ231543.1 |
- |
5, 6 |
417 |
|
FQ231632.1 |
- |
5, 6 |
417 |
|
FQ231753.1 |
- |
5, 6 |
414 |
|
FQ231815.1 |
- |
5, 6 |
399 |
|
FQ231975.1 |
- |
5, 6 |
399 |
|
FQ232042.1 |
- |
5, 6 |
416 |
|
FQ232191.1 |
- |
5, 6 |
417 |
|
FQ232201.1 |
- |
5, 6 |
417 |
|
FQ232223.1 |
- |
5, 6 |
417 |
|
FQ232271.1 |
- |
5, 6 |
415 |
|
FQ232364.1 |
- |
5, 6 |
419 |
|
FQ232445.1 |
- |
5, 6 |
415 |
|
FQ232464.1 |
- |
5, 6 |
417 |
|
FQ232710.1 |
- |
5, 6 |
415 |
|
FQ232847.1 |
- |
5, 6 |
417 |
|
FQ232869.1 |
- |
5, 6 |
417 |
|
FQ232965.1 |
- |
5, 6 |
417 |
|
FQ232977.1 |
- |
5, 6 |
417 |
|
FQ232986.1 |
- |
5, 6 |
417 |
|
FQ233178.1 |
- |
5, 6 |
417 |
|
FQ233283.1 |
- |
5, 6 |
396 |
|
FQ233669.1 |
- |
5, 6 |
416 |
|
FQ233698.1 |
- |
5, 6 |
399 |
|
FQ233777.1 |
- |
5, 6 |
415 |
|
FQ233810.1 |
- |
5, 6 |
419 |
|
FQ233845.1 |
- |
5, 6 |
417 |
|
FQ233850.1 |
- |
5, 6 |
418 |
|
FQ233926.1 |
- |
5, 6 |
417 |
|
FQ234037.1 |
- |
5, 6 |
417 |
|
FQ234069.1 |
- |
5, 6 |
417 |
|
FQ234108.1 |
- |
5, 6 |
417 |
|
FQ234199.1 |
- |
5, 6 |
417 |
|
FQ234410.1 |
- |
5, 6 |
399 |
|
FQ234418.1 |
- |
5, 6 |
417 |
|
FQ234523.1 |
- |
5, 6 |
404 |
|
FQ234589.1 |
- |
5, 6 |
415 |
|
FQ234657.1 |
- |
5, 6 |
399 |
|
FQ234685.1 |
- |
5, 6 |
417 |
|
FQ234766.1 |
- |
5, 6 |
414 |
|
FQ235008.1 |
- |
5, 6 |
419 |
|
FQ235038.1 |
- |
5, 6 |
417 |
|
FQ235049.1 |
- |
5, 6 |
399 |
|
Y13714.1 |
- |
6, 7 |
383 |
|
View Full Product Details