Gene Transcripts
- Gene Symbol
- Psap
- Entrez Gene ID
- 25524
- Gene Name
- prosaposin
- Gene Aliases
- SGP-1, SGP1A
- Chromosome Location
- Chr. 20: 29831302 - 29856876 on Build Rnor_6.0
- UniGene ID
- Rn.97173
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190236.1 |
NP_001177165.1
|
3, 4 |
334 |
|
NM_001190237.1 |
NP_001177166.1
|
3, 4 |
334 |
|
NM_001190238.1 |
NP_001177167.1
|
3, 4 |
334 |
|
NM_013013.2 |
NP_037145.2
|
3, 4 |
334 |
|
XM_006256403.1 |
XP_006256465.1
|
3, 4 |
346 |
|
GenBank mRNA |
BC061759.1 |
- |
3, 4 |
268 |
|
CK365248.1 |
- |
3, 4 |
334 |
|
CO403947.1 |
- |
1, 2 |
102 |
|
FQ217536.1 |
- |
3, 4 |
285 |
|
M19936.1 |
- |
3, 4 |
284 |
|
S81353.1 |
- |
3, 4 |
284 |
|
View Full Product Details