Gene Transcripts

Gene Symbol
Ttpa
Entrez Gene ID
25571
Gene Name
tocopherol (alpha) transfer protein
Gene Aliases
TTP
Chromosome Location
Chr. 5: 34007769 - 34029315 on Build Rnor_6.0
UniGene ID
Rn.228762
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013048.2 NP_037180.1 5 784
XM_008763588.1 XP_008761810.1 7 791
XM_008763589.1 XP_008761811.1 6 734

View Full Product Details