Gene Transcripts

Gene Symbol
Cd37
Entrez Gene ID
29185
Gene Name
CD37 molecule
Gene Aliases
Chromosome Location
Chr. 1: 101230470 - 101236050 on Build Rnor_6.0
UniGene ID
Rn.2357
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017124.1 NP_058820.1 2 144
XM_006228981.2 XP_006229043.1 3 337
XM_008759326.1 XP_008757548.1 2 187
GenBank mRNA BC059144.1 - 2 185
FQ230385.1 - 2 162
X53517.1 - 2 144

View Full Product Details