Gene Transcripts

Gene Symbol
Mpz
Entrez Gene ID
24564
Gene Name
myelin protein zero
Gene Aliases
MPP, P0
Chromosome Location
Chr. 13: 89524349 - 89529248 on Build Rnor_6.0
UniGene ID
Rn.8997
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017027.1 NP_058723.1 2 253
GenBank mRNA BC078859.1 - 253
K03242.1 - 2 253

View Full Product Details