Gene Transcripts
- Gene Symbol
- TPST2
- Entrez Gene ID
- 8459
- Gene Name
- tyrosylprotein sulfotransferase 2
- Gene Aliases
- TANGO13B
- Chromosome Location
- Chr. 22: 26524082 - 26590125 on Build GRCh38
- UniGene ID
- Hs.632768
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001008566.1 |
NP_001008566.1
|
5, 6 |
1430 |
|
NM_003595.3 |
NP_003586.3
|
5, 6 |
1322 |
|
XM_006724338.3 |
XP_006724401.1
|
6, 7 |
1441 |
|
GenBank mRNA |
AF049891.1 |
- |
5, 6 |
1286 |
|
AF061254.1 |
- |
3, 4 |
1094 |
|
AJ006198.1 |
- |
3, 4 |
1176 |
|
AK074538.1 |
- |
5, 6 |
1322 |
|
AK075139.1 |
- |
5, 6 |
1394 |
|
AK304545.1 |
- |
7, 8 |
1406 |
|
AL136623.2 |
- |
5, 6 |
1306 |
|
BC001057.2 |
- |
5, 6 |
1301 |
3509627 |
BC017509.1 |
- |
5, 6 |
1286 |
4857366 |
CR456597.1 |
- |
3, 4 |
1184 |
|
CR533528.1 |
- |
3, 4 |
1089 |
|
View Full Product Details