Gene Transcripts

Gene Symbol
Apcs
Entrez Gene ID
29339
Gene Name
amyloid P component, serum
Gene Aliases
Sap
Chromosome Location
Chr. 13: 91426502 - 91427988 on Build Rnor_6.0
UniGene ID
Rn.109007
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017170.2 NP_058866.2 2 495
XM_008769786.1 XP_008768008.1 3 804
GenBank mRNA M83177.1 - 2 495
X55761.1 - 2 577

View Full Product Details