Gene Transcripts

Gene Symbol
Esp31
Entrez Gene ID
100126768
Gene Name
exocrine gland secreted peptide 31
Gene Aliases
Gm20516
Chromosome Location
Chr. 17: 38639447 - 38645653 on Build GRCm38
UniGene ID
Mm.471528
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177586.1 NP_001171057.1 3 259
GenBank mRNA AB307010.1 - 3 259

View Full Product Details