Gene Transcripts

Gene Symbol
ND4L
Entrez Gene ID
17720
Gene Name
NADH dehydrogenase subunit 4L
Gene Aliases
Chromosome Location
Chr. : - on Build
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK140187.1 - 1 265
AK144853.1 - 265
AK153747.1 - 264
AK157367.1 - 265

View Full Product Details