Gene Transcripts

Gene Symbol
Akna
Entrez Gene ID
100182
Gene Name
AT-hook transcription factor
Gene Aliases
AI597013
Chromosome Location
Chr. 4: 63367127 - 63410758 on Build GRCm38
UniGene ID
Mm.387725
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK089194.1 - 809
AK129468.1 - 801
AK154663.1 - 1 20
AK158313.1 - 3 713
AK159547.1 - 884
AK170121.1 - 1 20
AK170885.1 - 776
BC092227.1 - 2 665 30531788

View Full Product Details