Gene Transcripts

Gene Symbol
Vmn1r88
Entrez Gene ID
100312474
Gene Name
vomeronasal 1 receptor, 88
Gene Aliases
Chromosome Location
Chr. 7: 13177719 - 13178669 on Build GRCm38
UniGene ID
Mm.480295
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167537.1 NP_001161009.1 1 178

View Full Product Details