Gene Transcripts

Gene Symbol
Gm17365
Entrez Gene ID
100312956
Gene Name
predicted gene, 17365
Gene Aliases
Pate-dj, Pate3
Chromosome Location
Chr. 9: 35645113 - 35648202 on Build GRCm38
UniGene ID
Mm.475850
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167592.2 NP_001161064.1 3 996
GenBank mRNA EU004071.1 - 3 996

View Full Product Details