Gene Transcripts
- Gene Symbol
- Gm17727
- Entrez Gene ID
- 100312986
- Gene Name
- predicted gene, 17727
- Gene Aliases
- Pate-e
- Chromosome Location
- Chr. 9: 35776524 - 35778109 on Build GRCm38
- UniGene ID
- Mm.437659
- Species
- Mus musculus
- Gene Symbol
- D730048I06Rik
- Entrez Gene ID
- 68171
- Gene Name
- RIKEN cDNA D730048I06 gene
- Gene Aliases
- Pate-C, mANLP2
- Chromosome Location
- Chr. 9: 35788050 - 35790112 on Build GRCm38
- UniGene ID
- Mm.99495
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK136605.1 |
- |
|
772 |
|
View Full Product Details