Gene Transcripts

Gene Symbol
Dthd1
Entrez Gene ID
100322896
Gene Name
death domain containing 1
Gene Aliases
Gm17384
Chromosome Location
Chr. 5: 62813823 - 62888318 on Build GRCm38
UniGene ID
Mm.479821
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170705.1 NP_001164176.1 1 287

View Full Product Details