Gene Transcripts

Gene Symbol
Ttc7b
Entrez Gene ID
104718
Gene Name
tetratricopeptide repeat domain 7B
Gene Aliases
AA408451, Ttc7l1
Chromosome Location
Chr. 12: 100300770 - 100520822 on Build GRCm38
UniGene ID
Mm.295441
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033213.1 NP_001028385.1 4, 5 711
GenBank mRNA AK046740.1 - 4, 5 680
AK148363.1 - 4, 5 683

View Full Product Details