Gene Transcripts
- Gene Symbol
- Brd9
- Entrez Gene ID
- 105246
- Gene Name
- bromodomain containing 9
- Gene Aliases
- AL022779
- Chromosome Location
- Chr. 13: 73937825 - 73960895 on Build GRCm38
- UniGene ID
- Mm.278509
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC031484.1 |
- |
16 |
2340 |
|
BC044769.1 |
- |
9 |
2283 |
|
BC089379.1 |
- |
16 |
2297 |
|
View Full Product Details