Gene Transcripts

Gene Symbol
Btaf1
Entrez Gene ID
107182
Gene Name
BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, (Mot1 homolog, S. cerevisiae)
Gene Aliases
AI414500, AI447930, E430027O22Rik, TAF170
Chromosome Location
Chr. 19: 36926079 - 37014057 on Build GRCm38
UniGene ID
Mm.295062
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080706.1 NP_001074175.1 6 988
GenBank mRNA AB287295.1 - 6 713

View Full Product Details