Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017233.1 | NP_058929.1 | 4 | 212 | |
GenBank mRNA | AF082834.1 | - | 4 | 212 | |
BC081819.1 | - | 4 | 210 | 7107086 | |
FQ209393.1 | - | 4 | 220 | ||
FQ209489.1 | - | 3 | 172 | ||
FQ210905.1 | - | 4 | 220 | ||
FQ218378.1 | - | 4 | 220 | ||
FQ218745.1 | - | 4 | 219 | ||
FQ219696.1 | - | 4 | 220 | ||
M18405.1 | - | 3 | 168 |