Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027773.3 | NP_082049.1 | 3 | 368 | |
GenBank mRNA | AK077374.1 | - | 3 | 329 | |
AK160183.1 | - | 3 | 329 | ||
BC115519.1 | - | 3 | 367 | 40101189 | |
BC115520.1 | - | 3 | 368 | 40101190 |