Gene Transcripts
- Gene Symbol
- Klra9
- Entrez Gene ID
- 16640
- Gene Name
- killer cell lectin-like receptor subfamily A, member 9
- Gene Aliases
- LY49I1, Ly49I
- Chromosome Location
- Chr. 6: 130178626 - 130194374 on Build GRCm38
- UniGene ID
- Mm.457996
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010651.3 |
NP_034781.2
|
8 |
1111 |
|
View Full Product Details